ADV371425460 AChemBlock

3-Amino-5-bromo-1H-pyrazolo[3,4-b]pyrazine

  • MDL number MFCD17016134
  • Cas Number : 1242336-77-1
  • Molecular Weight : 214.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-amino-5-bromo-1h-pyrazolo-3-4-b-pyrazine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3368
pictograms GHS07
hstatements H313;H303;H333
Availability details for
Quantity
  • Availability
  • Add to Cart