ADV371425461 AChemBlock

1-(5-Fluoropyridin-2-yl)ethanamine

  • MDL number MFCD18260255
  • Cas Number : 915720-57-9
  • Molecular Weight : 140.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellowish liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-5-fluoropyridin-2-yl-ethanamine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3369
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart