ADV371425465 AChemBlock

6-Amino-2,3-dihydro-2-methyl-1H-isoindol-1-one

  • MDL number MFCD13178503
  • Cas Number : 69189-26-0
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-amino-2-3-dihydro-2-methyl-1h-isoindol-1-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3374
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart