ADV371425466 AChemBlock

6-Amino-1-methyl-2-oxoindoline

  • MDL number MFCD17015857
  • Cas Number : 813424-16-7
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid/pale white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-amino-1-methyl-2-oxoindoline.html

Safety Information

signalword Warning
pcodes P260;P264;P270;P271;P280
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3375
pictograms GHS07
hstatements H313;H303;H333
Availability details for
Quantity
  • Availability
  • Add to Cart