ADV371425470 AChemBlock

2-Chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine

  • MDL number MFCD13189363
  • Cas Number : 1060815-90-8
  • Molecular Weight : 279.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-5-iodo-7h-pyrrolo-2-3-d-pyrimidine.html

Safety Information

signalword Danger
pcodes P280;P310;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3379
pictograms GHS05; GHS07
hstatements H302;H314;H318
Availability details for
Quantity
  • Availability
  • Add to Cart