ADV371425471 AChemBlock

5-Amino-2,3-dihydro-2-methyl-1H-isoindol-1-one

  • MDL number MFCD17214360
  • Cas Number : 1190380-38-1
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-amino-2-3-dihydro-2-methyl-1h-isoindol-1-one.html

Safety Information

signalword Danger
pcodes P264;P280;P301+330+331;P303+361+353;P304+340;P305+351+338;P310;P363
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3380
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart