ADV371425483 AChemBlock

Methyl 4-bromo-6-(1H)-indazole carboxylate

  • MDL number MFCD07368212
  • Cas Number : 885518-47-8
  • Molecular Weight : 255.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-bromo-6-1h-indazole-carboxylate.html

Safety Information

signalword Danger
pcodes P261;P264;P270;P271;P280;P301+312;P304+340;P305+351+338;P310;P332+313;P403+233;
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3393
pictograms GHS05; GHS07
hstatements H302;H332;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart