ADV371425487 AChemBlock

3-Chloropyrazine-2-carbonitrile

  • MDL number MFCD00219653
  • Cas Number : 55557-52-3
  • Molecular Weight : 139.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form brown crystalline powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-chloropyrazine-2-carbonitrile.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3397
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart