ADV371425489 AChemBlock

2-Oxopiperidine-4-methanol

  • MDL number MFCD17016089
  • Cas Number : 53611-47-5
  • Molecular Weight : 129.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid/white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-oxopiperidine-4-methanol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3400
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart