ADV371425490 AChemBlock

5-Amino-piperidin-2-one hydrochloride

  • MDL number MFCD09743647
  • Cas Number : 1235440-18-2
  • Molecular Weight : 150.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-amino-piperidin-2-one-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3401
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart