ADV371425498 AChemBlock

1-Boc-3-(cyanomethylene)azetidine

  • MDL number MFCD12761241
  • Cas Number : 1153949-11-1
  • Molecular Weight : 194.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-cyanomethylene-azetidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3409
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart