ADV371425501 AChemBlock

4-(Iodomethyl)tetrahydropyran

  • MDL number MFCD06410724
  • Cas Number : 101691-94-5
  • Molecular Weight : 226.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-iodomethyl-tetrahydropyran.html

Safety Information

signalword Warning
pcodes P261;P264;P270;P271;P280;P301+312;P302+352;P304+340;P305+351+338;P337+313;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3412
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart