ADV371425502 AChemBlock

4-(Aminomethyl)tetrahydropyran

  • MDL number MFCD02179435
  • Cas Number : 130290-79-8
  • Molecular Weight : 115.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-aminomethyl-tetrahydropyran.html

Safety Information

signalword Warning
pcodes P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3413
pictograms GHS07
hstatements H302;H317;H320
Availability details for
Quantity
  • Availability
  • Add to Cart