ADV371425512 AChemBlock

5-Amino-1,3,3-trimethyl-2-oxindole

  • MDL number MFCD16994189
  • Cas Number : 953048-71-0
  • Molecular Weight : 190.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellowish solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-amino-1-3-3-trimethyl-2-oxindole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3424
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart