ADV371425521 AChemBlock

3-Amino-pyrazine-2-carbaldehyde

  • MDL number MFCD10696886
  • Cas Number : 32710-14-8
  • Molecular Weight : 123.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-amino-pyrazine-2-carbaldehyde.html

Safety Information

signalword Danger
pcodes P260;P264;P270;P280;P301+312;P301+330+331;P303+361+353;P304+340;P305+351+338;P310;P363
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3433
pictograms GHS05; GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart