ADV371425526 AChemBlock

3-Methylamino-5-bromo-1H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD18432771
  • Cas Number : 1289189-96-3
  • Molecular Weight : 227.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form pink solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-methylamino-5-bromo-1h-pyrazolo-3-4-b-pyridine.html

Safety Information

signalword Warning
pcodes P261;P264;P270;P280;P301+312;P302+352;P304+340;P305+351+338;P337+313;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3439
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart