ADV371425530 AChemBlock

2-Chloro-5H-pyrrolo[2,3-b]pyrazine

  • MDL number MFCD11518976
  • Cas Number : 889447-19-2
  • Molecular Weight : 153.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white or brown solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-5h-pyrrolo-2-3-b-pyrazine.html

Safety Information

signalword Warning
pcodes P261;P264;P270;P271;P280;P301+312;P302+352;P304+340;P305+351+338;P337+313;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3444
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart