ADV371425536 AChemBlock

4-Chloro-6-azaindole

  • MDL number MFCD16657181
  • Cas Number : 1188313-15-6
  • Molecular Weight : 152.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-6-azaindole.html

Safety Information

signalword Warning
pcodes P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3450
pictograms GHS07
hstatements H303;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart