ADV371425537 AChemBlock

5-Bromo-3-nitropyridine-2-carbaldehyde

  • MDL number MFCD17016035
  • Cas Number : 1086838-46-1
  • Molecular Weight : 231.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form White powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-3-nitropyridine-2-carbaldehyde.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3451
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart