ADV371425539 AChemBlock

3-Methyl-1H-pyrazolo[4,3-b]pyridine

  • MDL number MFCD08062968
  • Cas Number : 194278-45-0
  • Molecular Weight : 133.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-methyl-1h-pyrazolo-4-3-b-pyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3453
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart