ADV371425545 AChemBlock

8-Amino-3-bromo-imidazo[1,2-a]pyridine

  • MDL number MFCD17012030
  • Cas Number : 1232431-81-0
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light gray solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/8-amino-3-bromo-imidazo-1-2-a-pyridine.html

Safety Information

signalword Warning
pcodes P261;P264;P270;P271;P280;P301+312;P302+352;P304+340;P305+351+338;P337+313;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3460
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart