ADV371425548 AChemBlock

5-Ethynyl-2-methoxypyridine

  • MDL number MFCD10698135
  • Cas Number : 663955-59-7
  • Molecular Weight : 133.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-ethynyl-2-methoxypyridine.html

Safety Information

signalword Warning
pcodes P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3464
pictograms GHS07
hstatements H320
Availability details for
Quantity
  • Availability
  • Add to Cart