ADV371425550 AChemBlock

5-Fluoro-2-hydroxymethylpyridine

  • MDL number MFCD09027049
  • Cas Number : 802325-29-7
  • Molecular Weight : 127.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow liquid/light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-fluoro-2-hydroxymethylpyridine.html

Safety Information

signalword Danger
pcodes P264;P270;P280;P301+312;P305+351+338;P310
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3467
pictograms GHS05; GHS07
hstatements H302;H318
Availability details for
Quantity
  • Availability
  • Add to Cart