ADV371425551 AChemBlock

3-Fluoro-4-(4-methyl-1-piperazinyl)benzenamine

  • MDL number MFCD05663793
  • Cas Number : 221198-99-8
  • Molecular Weight : 209.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-fluoro-4-4-methyl-1-piperazinyl-benzenamine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3469
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart