ADV371425558 AChemBlock

2,8-Diazaspiro[4.5]decan-3-one

  • MDL number MFCD11044206
  • Cas Number : 561314-57-6
  • Molecular Weight : 154.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-8-diazaspiro-4-5-decan-3-one.html

Safety Information

signalword Danger
pcodes P201;P261;P280;P310;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3479
pictograms GHS02; GHS05
hstatements H226;H314;H318
Availability details for
Quantity
  • Availability
  • Add to Cart