ADV371425562 AChemBlock

7-Azabicyclo[2.2.1]heptane hydrochloride

  • MDL number MFCD00154183
  • Cas Number : 27514-07-4
  • Molecular Weight : 133.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-azabicyclo-2-2-1-heptane-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3483
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart