ADV371425566 AChemBlock

3-Bromomethyl-3-methyloxetane

  • MDL number MFCD02684290
  • Cas Number : 78385-26-9
  • Molecular Weight : 165.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-bromomethyl-3-methyloxetane.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3487
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart