ADV371425567 AChemBlock

2-Methyl-N-(oxetan-3-ylidene)propane-2-sulfinamide

  • MDL number MFCD14702522
  • Cas Number : 1158098-73-7
  • Molecular Weight : 175.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-methyl-n-oxetan-3-ylidene-propane-2-sulfinamide.html

Safety Information

signalword Warning
pcodes P261;P264;P271;P280;P302+352;P304+340;P305+351+338;P337+313;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3488
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart