ADV371425577 AChemBlock

(5-Fluoropyridin-3-yl)methanamine

  • MDL number MFCD08450424
  • Cas Number : 23586-96-1
  • Molecular Weight : 126.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-fluoropyridin-3-yl-methanamine.html

Safety Information

signalword Danger
pcodes P261;P264;P270;P271;P280;P301+310;P302;P304+340;P305+351+338;P310;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3498
pictograms GHS06; GHS05
hstatements H301;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart