ADV371425582 AChemBlock

5-Amino-5,6,7,8-tetrahydroquinoline

  • MDL number MFCD04038438
  • Cas Number : 71569-15-8
  • Molecular Weight : 148.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-amino-5-6-7-8-tetrahydroquinoline.html

Safety Information

signalword Warning
pcodes P261;P264;P271;P280;P302+352;P304+340;P305+351+338;P337+313;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3503
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart