ADV371425584 AChemBlock

4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carbaldehyde

  • MDL number MFCD12406099
  • Cas Number : 908287-21-8
  • Molecular Weight : 181.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-7h-pyrrolo-2-3-d-pyrimidine-5-carbaldehyde.html

Safety Information

signalword Warning
pcodes P210;P261;P264;P271;P280;P302+352;P304+340;P305+351+338;P337+313;P370+378;P403+233;P403+235
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3505
pictograms GHS07
hstatements H227;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart