ADV371425588 AChemBlock

3-Bromo-4,7-diazaindole

  • MDL number MFCD09834812
  • Cas Number : 56015-31-7
  • Molecular Weight : 198.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form earth yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-4-7-diazaindole.html

Safety Information

signalword Danger
pcodes P210;P280;P305+351+338;P310;P370+378;P403+235
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3510
pictograms GHS05
hstatements H227;H318
Availability details for
Quantity
  • Availability
  • Add to Cart