ADV371425589 AChemBlock

5-Amino-7-fluoro-1H-indole

  • MDL number MFCD12923890
  • Cas Number : 926028-84-4
  • Molecular Weight : 150.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-amino-7-fluoro-1h-indole.html

Safety Information

signalword warning
pcodes P210;P233;P240;P241;P242;P243;P264;P280;P303+361+353;P370+378;P403+235
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3511
pictograms GHS05
hstatements H226
Availability details for
Quantity
  • Availability
  • Add to Cart