ADV371425591 AChemBlock

7-Chloro-furo[3,2-b]pyridine

  • MDL number MFCD15143870
  • Cas Number : 182691-75-4
  • Molecular Weight : 153.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form gray solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-chloro-furo-3-2-b-pyridine.html

Safety Information

signalword Warning
pcodes P273;P391
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3513
pictograms GHS09
hstatements H410
Availability details for
Quantity
  • Availability
  • Add to Cart