ADV371425602 AChemBlock

Thieno[3,2-d]pyrimidine-4-carboxylic acid

  • MDL number MFCD17015875
  • Cas Number : 1029144-49-7
  • Molecular Weight : 180.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/thieno-3-2-d-pyrimidine-4-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P264;P270;P301+312
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3525
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart