ADV371425603 AChemBlock

tert-butyl 3-(2-methoxy-2-oxoethyl)pyrrolidine-1-carboxylate

  • MDL number MFCD18910910
  • Cas Number : 890849-27-1
  • Molecular Weight : 243.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Store at Room Tem.
Shipping Temperature Store at Room Tem.
Country of Origin CN
Product Link https://www.achemblock.com/3-pyrrolidineacetic-acid-1-1-1-dimethylethoxy-carbonyl-methyl-ester.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3526
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart