ADV371425607 AChemBlock

5-Bromo[1,2,4]triazolo[4,3-a]pyridine

  • MDL number MFCD11043995
  • Cas Number : 1172085-67-4
  • Molecular Weight : 198.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-1-2-4-triazolo-4-3-a-pyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3531
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart