ADV371425617 AChemBlock

2-Boc-7,9-dioxo-2,6-diazaspiro[3.5]nonane

  • MDL number MFCD13184430
  • Cas Number : 1105664-04-7
  • Molecular Weight : 254.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-boc-7-9-dioxo-2-6-diazaspiro-3-5-nonane.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3543
pictograms GHS07
hstatements H303
Availability details for
Quantity
  • Availability
  • Add to Cart