ADV371425618 AChemBlock

Diethyl 1-Boc-azetidine-3-dicarboxylate

  • MDL number MFCD20126212
  • Cas Number : 1011479-75-6
  • Molecular Weight : 301.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow liquid/colorless liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/diethyl-1-boc-azetidine-3-dicarboxylate.html

Safety Information

signalword Danger
pcodes P301+310;
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3544
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart