ADV371425620 AChemBlock

Diethyl 1-benzylazetidine-3,3-dicarboxylate

  • MDL number MFCD17169917
  • Cas Number : 642411-11-8
  • Molecular Weight : 291.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/diethyl-1-benzylazetidine-3-3-dicarboxylate.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3546
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart