ADV371425621 AChemBlock

1-Cbz-3-azetidinone

  • MDL number MFCD09954923
  • Cas Number : 105258-93-3
  • Molecular Weight : 205.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white crystalline solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-cbz-3-azetidinone.html

Safety Information

signalword Danger
pcodes P261;P264;P270;P271;P280;P301+312;P302+352;P304+340;P305+351+338;P310;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3547
pictograms GHS05; GHS07
hstatements H302;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart