ADV371425622 AChemBlock

1-Cbz-3-(aminomethyl)azetidine

  • MDL number MFCD07772065
  • Cas Number : 1016731-24-0
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-cbz-3-aminomethyl-azetidine.html

Safety Information

signalword warning
pcodes P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3548
pictograms GHS07
hstatements H302;H320
Availability details for
Quantity
  • Availability
  • Add to Cart