ADV371425630 AChemBlock

Methyl 1-Cbz-azetidine-3-carboxylate

  • MDL number MFCD11110708
  • Cas Number : 757239-60-4
  • Molecular Weight : 249.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid/colorless or light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-1-cbz-azetidine-3-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3556
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart