ADV371425647 AChemBlock

tert-Butyl 2-azaspiro[3.3]hept-6-ylcarbamate

  • MDL number MFCD11858159
  • Cas Number : 1118786-85-8
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-azaspiro-3-3-hept-6-ylcarbamate.html

Safety Information

signalword Danger
pcodes P264;P270;P301+310;
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3575
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart