ADV371425648 AChemBlock

2-Cbz-2-aza-spiro[3.3]heptane-6-carboxylic acid

  • MDL number MFCD18432778
  • Cas Number : 1291487-33-6
  • Molecular Weight : 275.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-cbz-2-aza-spiro-3-3-heptane-6-carboxylic-acid.html

Safety Information

signalword Danger
pcodes P210;P261;P264;P270;P271;P280;P304+340;P305+351+338;P310;P403+233;P403+235
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3576
pictograms GHS05; GHS07
hstatements H227;H302;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart