ADV371425650 AChemBlock

2-(Boc-amino)-6-oxospiro[3.3]heptane

  • MDL number MFCD11858160
  • Cas Number : 1118786-86-9
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white solid/white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-boc-amino-6-oxospiro-3-3-heptane.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3578
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart