ADV371425654 AChemBlock

6-(Methoxycarbonyl)spiro[3.3]heptane-2-carboxylic acid

  • MDL number MFCD13195611
  • Cas Number : 10481-25-1
  • Molecular Weight : 198.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-methoxycarbonyl-spiro-3-3-heptane-2-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P264;P271;P280;P302+352;P304+340;P305+351+338;P337+313;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3582
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart