ADV371425656 AChemBlock

Ethyl 1-Boc-4-piperidone-3-carboxylate

  • MDL number MFCD01862189
  • Cas Number : 98977-34-5
  • Molecular Weight : 271.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-1-boc-4-piperidone-3-carboxylate.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3585
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart