ADV371425657 AChemBlock

1-Cbz-3-piperidone

  • MDL number MFCD03001723
  • Cas Number : 61995-20-8
  • Molecular Weight : 233.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form orange liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-cbz-3-piperidone.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3586
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart