ADV371425658 AChemBlock

4-Chloro-6-methyl-7H-pyrrolo[2,3-d]pyrimidine

  • MDL number MFCD09033903
  • Cas Number : 35808-68-5
  • Molecular Weight : 167.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-6-methyl-7h-pyrrolo-2-3-d-pyrimidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3587
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart